Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181820.2 | NP_861541.2 | 12 - 13 | 1860 | 61 | |
GenBank mRNA | AK050105.1 | - | 11 - 12 | 1755 | 61 | |
AY236493.1 | - | 12 - 13 | 1823 | 61 | ||
AY263162.1 | - | 12 - 13 | 1831 | 61 | ||
BC022758.1 | - | 4 - 5 | 550 | 61 | ||
BC040466.1 | - | 12 - 13 | 1772 | 61 | ||
CN719678.1 | - | 1 - 2 | 54 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |