Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183178.2 | NP_899001.1 | 9 - 10 | 1088 | 68 | |
GenBank mRNA | AK140132.1 | - | 9 - 10 | 1122 | 68 | |
AK144257.1 | - | 9 - 10 | 1023 | 68 | ||
BC055106.1 | - | 9 - 10 | 1088 | 6532001 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |