Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183278.2 | NP_899101.1 | 1 - 2 | 109 | 55 | |
GenBank mRNA | AK008614.1 | - | 1 - 2 | 109 | 55 | |
BC086894.1 | - | 1 - 2 | 111 | 55 | ||
BC147104.1 | - | 1 - 2 | 98 | 9056159 | 55 | |
BC147105.1 | - | 1 - 2 | 98 | 9056136 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |