Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081038.2 | NP_001074507.2 | 1 - 2 | 307 | 99 | |
GenBank mRNA | AK087912.1 | - | 1 - 2 | 308 | 99 | |
BB666983.1 | - | 1 - 2 | 308 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |