Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081038.2 | NP_001074507.2 | 6 - 7 | 868 | 113 | |
GenBank mRNA | AK087912.1 | - | 6 - 7 | 870 | 113 | |
BB559682.2 | - | 4 - 5 | 427 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |