Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143683.1 | NP_001137155.1 | 3 - 4 | 791 | 90 | |
NM_029837.1 | NP_084113.1 | 3 - 4 | 504 | 90 | ||
GenBank mRNA | AK012553.1 | - | 3 - 4 | 505 | 90 | |
AK161666.1 | - | 3 - 4 | 792 | 90 | ||
AK165186.1 | - | 3 - 4 | 988 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |