Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045569.1 | 5 - 6 | 675 | 70 | ||
NR_103722.1 | 4 - 5 | 635 | 70 | |||
GenBank mRNA | AF364070.1 | - | 4 - 5 | 474 | 70 | |
AJ245772.1 | - | 4 - 5 | 490 | 70 | ||
AK003105.1 | - | 4 - 5 | 482 | 70 | ||
AK010172.1 | - | 4 - 5 | 482 | 70 | ||
AY026524.1 | - | 4 - 5 | 481 | 70 | ||
BC051197.1 | - | 4 - 5 | 434 | 1397202 | 70 | |
BC061051.1 | - | 4 - 5 | 396 | 30300606 | 70 | |
BC132358.1 | - | 4 - 5 | 310 | 40130635 | 70 | |
BC138801.1 | - | 4 - 5 | 310 | 8861823 | 70 | |
BQ176246.1 | - | 2 - 3 | 70 | |||
W18646.1 | - | 5 - 6 | 388 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |