Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161483.1 | NP_001154955.1 | 6 - 7 | 896 | 106 | |
NM_199024.2 | NP_950189.2 | 6 - 7 | 2666 | 106 | ||
GenBank mRNA | AK029800.1 | - | 2 - 3 | 400 | 106 | |
AK031008.1 | - | 6 - 7 | 2666 | 106 | ||
AK086574.1 | - | 2 - 3 | 400 | 106 | ||
BC056377.1 | - | 6 - 7 | 889 | 6850243 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |