Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013505.3 | NP_038533.1 | 14 - 15 | 2509 | 66 | |
GenBank mRNA | AK028604.1 | - | 15 - 16 | 2549 | 66 | |
AK028636.1 | - | 14 - 15 | 2510 | 66 | ||
AK030333.1 | - | 14 - 15 | 2510 | 66 | ||
AK048547.1 | - | 14 - 15 | 2510 | 66 | ||
AK078887.1 | - | 2 - 3 | 200 | 66 | ||
AK172168.1 | - | 14 - 15 | 2344 | 66 | ||
BC004663.1 | - | 8 - 9 | 1396 | 66 | ||
BC057867.1 | - | 14 - 15 | 2289 | 5327748 | 66 | |
L33779.1 | - | 14 - 15 | 2308 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |