Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004365.1 | NP_001004365.1 | 10 - 11 | 1136 | 95 | |
GenBank mRNA | AK035798.1 | - | 9 - 10 | 1048 | 95 | |
BC053106.1 | - | 9 - 10 | 1109 | 95 | ||
BC082279.1 | - | 10 - 11 | 1136 | 6332588 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |