Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
NM_001177960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010782.mm.1 |
TC0X00001682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
NM_001177960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010784.mm.1 |
TC0X00001682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177957.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177957.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177958.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177958.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177959.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177959.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
NM_001177960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010783.mm.1 |
TC0X00001682.mm.1 |
MTA TC |