Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NR_033143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
NR_033143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010555.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NR_033143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
NR_033143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010551.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NR_033143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
NR_033143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010554.mm.1 |
TC0X00001653.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_001168568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_001168568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_001168569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_001168569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_001168571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_001168571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_018737.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NM_018737.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NR_033143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
NR_033143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00010549.mm.1 |
TC0X00001653.mm.1 |
MTA TC |