Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244617.1 | NP_001231546.1 | 3 - 4 | 749 | 80 | |
NM_001244618.1 | NP_001231547.1 | 2 - 3 | 558 | 80 | ||
NM_008080.5 | NP_032106.1 | 3 - 4 | 749 | 80 | ||
NM_027739.2 | NP_082015.2 | 2 - 3 | 558 | 80 | ||
NR_045166.1 | 3 - 4 | 749 | 80 | |||
GenBank mRNA | AK040324.1 | - | 2 - 3 | 559 | 80 | |
AK086849.1 | - | 2 - 3 | 530 | 80 | ||
AK144484.1 | - | 3 - 4 | 715 | 80 | ||
BC019969.1 | - | 3 - 4 | 680 | 80 | ||
BC022180.1 | - | 3 - 4 | 537 | 80 | ||
BC036996.1 | - | 2 - 3 | 483 | 5038560 | 80 | |
BC057199.1 | - | 3 - 4 | 680 | 5315443 | 80 | |
L25885.1 | - | 2 - 3 | 512 | 80 | ||
U18975.1 | - | 2 - 3 | 387 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |