Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019429.2 | NP_062302.1 | 6 - 7 | 711 | 66 | |
GenBank mRNA | AJ131775.1 | - | 6 - 7 | 711 | 66 | |
AK088019.1 | - | 6 - 7 | 739 | 66 | ||
BC125481.1 | - | 6 - 7 | 709 | 40129996 | 66 | |
BC131955.1 | - | 6 - 7 | 709 | 40130232 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |