Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272064.1 | NP_001258993.1 | 16 - 17 | 2912 | 92 | |
NM_001272076.1 | NP_001259005.1 | 16 - 17 | 2594 | 92 | ||
NM_145076.4 | NP_659542.3 | 16 - 17 | 3014 | 92 | ||
GenBank mRNA | AB221637.1 | - | 16 - 17 | 2714 | 92 | |
AK133144.1 | - | 16 - 17 | 2594 | 92 | ||
AK166538.1 | - | 12 - 13 | 1971 | 92 | ||
BC025482.1 | - | 7 - 8 | 1109 | 92 | ||
BC056959.1 | - | 16 - 17 | 2992 | 5698079 | 92 | |
S78219.1 | - | 16 - 17 | 2912 | 92 | ||
S78221.1 | - | 16 - 17 | 3014 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |