Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163664.1 | NP_001157136.1 | 12 - 13 | 415 | 68 | |
NM_001163665.1 | NP_001157137.1 | 12 - 13 | 415 | 68 | ||
NM_001163666.1 | NP_001157138.1 | 10 - 11 | 388 | 68 | ||
NM_001163667.1 | NP_001157139.1 | 11 - 12 | 403 | 68 | ||
NM_001163668.1 | NP_001157140.1 | 10 - 11 | 364 | 68 | ||
NM_001163669.1 | NP_001157141.1 | 11 - 12 | 376 | 68 | ||
NM_001163670.1 | NP_001157142.1 | 8 - 9 | 331 | 68 | ||
NM_011620.3 | NP_035750.2 | 9 - 10 | 343 | 68 | ||
GenBank mRNA | AA028290.1 | - | 11 - 12 | 356 | 68 | |
AA771596.1 | - | 10 - 11 | 341 | 68 | ||
AI173874.1 | - | 10 - 11 | 380 | 68 | ||
AI574395.1 | - | 12 - 13 | 68 | |||
AI746973.1 | - | 10 - 11 | 383 | 68 | ||
BC003747.1 | - | 12 - 13 | 411 | 3594256 | 68 | |
CB196352.1 | - | 2 - 3 | 167 | 68 | ||
CK791812.1 | - | 1 - 2 | 53 | 68 | ||
DV049834.1 | - | 9 - 10 | 390 | 68 | ||
L48988.1 | - | 6 - 7 | 234 | 68 | ||
L48989.1 | - | 6 - 7 | 234 | 68 | ||
L48990.1 | - | 9 - 10 | 279 | 68 | ||
L48991.1 | - | 6 - 7 | 261 | 68 | ||
L48992.1 | - | 8 - 9 | 267 | 68 | ||
L48993.1 | - | 8 - 9 | 267 | 68 | ||
L49018.1 | - | 6 - 7 | 261 | 68 | ||
L49466.1 | - | 11 - 12 | 403 | 68 | ||
L49467.1 | - | 12 - 13 | 415 | 68 | ||
L49468.1 | - | 10 - 11 | 388 | 68 | ||
L49469.1 | - | 10 - 11 | 388 | 68 | ||
L49470.1 | - | 11 - 12 | 376 | 68 | ||
L49471.1 | - | 10 - 11 | 385 | 68 | ||
L49472.1 | - | 10 - 11 | 364 | 68 | ||
U77779.1 | - | 9 - 10 | 336 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |