Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013782.4 | NP_038810.2 | 8 - 9 | 902 | 64 | |
GenBank mRNA | AF099053.1 | - | 8 - 9 | 786 | 64 | |
AK036368.1 | - | 8 - 9 | 903 | 64 | ||
AK048026.1 | - | 8 - 9 | 885 | 64 | ||
AK081275.1 | - | 8 - 9 | 869 | 64 | ||
BC009013.1 | - | 6 - 7 | 569 | 64 | ||
BC053517.1 | - | 8 - 9 | 839 | 6315175 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |