Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301407.1 | NP_001288336.1 | 1 - 2 | 335 | 58 | |
NM_146173.3 | NP_666285.1 | 1 - 2 | 335 | 58 | ||
GenBank mRNA | AF276890.1 | - | 1 - 2 | 334 | 58 | |
AK004954.1 | - | 1 - 2 | 309 | 58 | ||
AK172176.1 | - | 1 - 2 | 316 | 58 | ||
BC024685.1 | - | 1 - 2 | 293 | 3981492 | 58 | |
BU054333.1 | - | 1 - 2 | 89 | 58 | ||
CJ068126.1 | - | 1 - 2 | 327 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |