Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143755.1 | NP_001137227.1 | 6 - 7 | 859 | 61 | |
NM_027164.1 | NP_081440.1 | 5 - 6 | 904 | 61 | ||
GenBank mRNA | AK006931.1 | - | 6 - 7 | 864 | 61 | |
BC050816.1 | - | 5 - 6 | 904 | 6774506 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |