Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201372.3 | NP_958760.2 | 3 - 4 | 397 | 76 | |
GenBank mRNA | AK052757.1 | - | 3 - 4 | 412 | 76 | |
AK162606.1 | - | 3 - 4 | 430 | 76 | ||
AK171754.1 | - | 404 | 76 | |||
BC055929.1 | - | 3 - 4 | 397 | 76 | ||
BC096560.1 | - | 3 - 4 | 402 | 1527981 | 76 | |
BC116877.1 | - | 3 - 4 | 411 | 40126196 | 76 | |
BC116879.1 | - | 3 - 4 | 411 | 40126198 | 76 | |
BC144712.1 | - | 3 - 4 | 411 | 76 | ||
BC144713.1 | - | 3 - 4 | 411 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |