Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029775.2 | NP_084051.1 | 5 - 6 | 676 | 119 | |
GenBank mRNA | AK012764.1 | - | 5 - 6 | 688 | 119 | |
AK013933.1 | - | 5 - 6 | 694 | 119 | ||
AK134862.1 | - | 723 | 119 | |||
BC020048.1 | - | 5 - 6 | 712 | 119 | ||
BC020089.1 | - | 5 - 6 | 676 | 4013755 | 119 | |
BC049641.1 | - | 865 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |