Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031397.2 | NP_113574.1 | 3 - 4 | 326 | 87 | |
GenBank mRNA | AF319464.1 | - | 3 - 4 | 327 | 87 | |
AK076154.1 | - | 3 - 4 | 379 | 87 | ||
AK089066.1 | - | 3 - 4 | 157 | 87 | ||
BC111523.2 | - | 3 - 4 | 327 | 40048106 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |