Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026085.2 | NP_080361.1 | 8 - 9 | 978 | 76 | |
GenBank mRNA | AK007784.1 | - | 7 - 8 | 733 | 76 | |
AK014198.1 | - | 8 - 9 | 979 | 76 | ||
AK018692.1 | - | 7 - 8 | 734 | 76 | ||
BC061073.1 | - | 7 - 8 | 733 | 30381600 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |