Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113355.1 | NP_001106826.1 | 3 - 4 | 444 | 119 | |
NM_133672.3 | NP_598433.1 | 3 - 4 | 371 | 119 | ||
GenBank mRNA | AK028096.1 | - | 3 - 4 | 445 | 119 | |
AK086851.1 | - | 267 | 119 | |||
AK147989.1 | - | 3 - 4 | 312 | 119 | ||
AK149859.1 | - | 3 - 4 | 296 | 119 | ||
AK150244.1 | - | 3 - 4 | 269 | 119 | ||
AK151360.1 | - | 3 - 4 | 281 | 119 | ||
AK152424.1 | - | 3 - 4 | 315 | 119 | ||
AK159783.1 | - | 3 - 4 | 321 | 119 | ||
AK165853.1 | - | 3 - 4 | 315 | 119 | ||
AK167592.1 | - | 3 - 4 | 326 | 119 | ||
AK168537.1 | - | 3 - 4 | 326 | 119 | ||
AK209699.1 | - | 2 - 3 | 234 | 119 | ||
AK216993.1 | - | 2 - 3 | 234 | 119 | ||
BC007148.1 | - | 3 - 4 | 292 | 3485844 | 119 | |
S41204.1 | - | 3 - 4 | 318 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |