Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900022879.mm.1 |
NM_027448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022879.mm.1 |
NM_027448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022879.mm.1 |
NM_029434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022879.mm.1 |
NM_029434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022879.mm.1 |
TC0900002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022880.mm.1 |
NM_027448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022880.mm.1 |
NM_027448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022880.mm.1 |
NM_029434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022880.mm.1 |
NM_029434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022880.mm.1 |
TC0900002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022881.mm.1 |
NM_027448.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022881.mm.1 |
NM_027448.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022881.mm.1 |
NM_029434.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022881.mm.1 |
NM_029434.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022881.mm.1 |
TC0900002766.mm.1 |
MTA TC |