Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 11 - 12 | 1531 | 63 | |
NM_027521.3 | NP_081797.1 | 10 - 11 | 1083 | 63 | ||
GenBank mRNA | AK044860.1 | - | 10 - 11 | 1045 | 63 | |
AK134029.1 | - | 4 - 5 | 505 | 63 | ||
AK151423.1 | - | 11 - 12 | 1475 | 63 | ||
AK159194.1 | - | 11 - 12 | 1532 | 63 | ||
AK159474.1 | - | 10 - 11 | 1575 | 63 | ||
AK161766.1 | - | 3 - 4 | 591 | 63 | ||
AK170262.1 | - | 11 - 12 | 1517 | 63 | ||
AK171304.1 | - | 11 - 12 | 1517 | 63 | ||
BC053750.1 | - | 10 - 11 | 1083 | 6333971 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |