Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033536.1 | NP_001028708.1 | 3 - 4 | 860 | 70 | |
GenBank mRNA | AK090370.1 | - | 3 - 4 | 861 | 70 | |
AK144999.1 | - | 4 - 5 | 692 | 70 | ||
AK170249.1 | - | 4 - 5 | 332 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |