Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183408.3 | NP_899668.1 | 6 - 7 | 984 | 97 | |
GenBank mRNA | AF038895.1 | - | 6 - 7 | 485 | 97 | |
AK153657.1 | - | 6 - 7 | 989 | 98 | ||
AK160843.1 | - | 5 - 6 | 418 | 97 | ||
BC027224.1 | - | 6 - 7 | 680 | 97 | ||
BC044786.2 | - | 5 - 6 | 1546 | 97 | ||
BC139405.1 | - | 6 - 7 | 824 | 8862427 | 97 | |
BC139408.1 | - | 6 - 7 | 824 | 8862430 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |