Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034895.3 | NP_001030067.2 | 3 - 4 | 398 | 97 | |
GenBank mRNA | AK165530.1 | - | 3 - 4 | 386 | 97 | |
BB641594.1 | - | 3 - 4 | 398 | 97 | ||
BC072571.1 | - | 3 - 4 | 374 | 97 | ||
CB249573.1 | - | 3 - 4 | 374 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |