Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083902.1 | NP_001077371.1 | 8 - 9 | 1554 | 81 | |
NM_134251.2 | NP_599012.2 | 9 - 10 | 1670 | 81 | ||
GenBank mRNA | AF319951.1 | - | 9 - 10 | 1664 | 81 | |
AK087781.1 | - | 5 - 6 | 1260 | 81 | ||
AK165080.1 | - | 4 - 5 | 1177 | 81 | ||
BC030926.1 | - | 8 - 9 | 1554 | 4910321 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |