Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018541.1 | - | 3 - 4 | 201 | 64 | |
BC057953.1 | - | 3 - 4 | 345 | 64 | ||
CB055593.1 | - | 3 - 4 | 323 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |