Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146048.1 | NP_001139520.1 | 11 - 12 | 1342 | 77 | |
NM_172528.3 | NP_766116.3 | 11 - 12 | 990 | 77 | ||
GenBank mRNA | AK147075.1 | - | 11 - 12 | 1342 | 77 | |
AK220166.1 | - | 11 - 12 | 1360 | 77 | ||
BC022613.1 | - | 3 - 4 | 284 | 77 | ||
BC046591.1 | - | 11 - 12 | 1329 | 77 | ||
BC087542.1 | - | 11 - 12 | 1189 | 4934980 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |