Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008404.4 | NP_032430.2 | 1 - 2 | 113 | 66 | |
GenBank mRNA | AK088435.1 | - | 1 - 2 | 102 | 66 | |
AK136502.1 | - | 1 - 2 | 114 | 66 | ||
AK161236.1 | - | 1 - 2 | 82 | 66 | ||
AK169587.1 | - | 1 - 2 | 88 | 66 | ||
X14951.1 | - | 1 - 2 | 79 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |