Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000015681.mm.1 |
NM_001252635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015681.mm.1 |
NM_001252635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015681.mm.1 |
NM_023370.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015681.mm.1 |
NM_023370.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015681.mm.1 |
NR_045556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015681.mm.1 |
NR_045556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015681.mm.1 |
TC1000002233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015686.mm.1 |
NM_001252635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015686.mm.1 |
NM_001252635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015686.mm.1 |
NM_023370.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015686.mm.1 |
NM_023370.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015686.mm.1 |
NR_045556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015686.mm.1 |
NR_045556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015686.mm.1 |
TC1000002233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015679.mm.1 |
NM_001252635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015679.mm.1 |
NM_001252635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015679.mm.1 |
NM_023370.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015679.mm.1 |
NM_023370.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015679.mm.1 |
NR_045556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015679.mm.1 |
NR_045556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015679.mm.1 |
TC1000002233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015680.mm.1 |
NM_001252635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015680.mm.1 |
NM_001252635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015680.mm.1 |
NM_023370.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015680.mm.1 |
NM_023370.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015680.mm.1 |
NR_045556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015680.mm.1 |
NR_045556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015680.mm.1 |
TC1000002233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015683.mm.1 |
NM_001252635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015683.mm.1 |
NM_001252635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015683.mm.1 |
NM_023370.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015683.mm.1 |
NM_023370.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015683.mm.1 |
NR_045556.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015683.mm.1 |
NR_045556.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015683.mm.1 |
TC1000002233.mm.1 |
MTA TC |