Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195268.1 | NP_001182197.1 | 1 - 2 | 106 | 72 | |
NR_036582.1 | 2 - 3 | 388 | 72 | |||
GenBank mRNA | AK137009.1 | - | 2 - 3 | 388 | 72 | |
BC052075.1 | - | 1 - 2 | 106 | 72 | ||
BM947270.1 | - | 2 - 3 | 388 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |