Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007585.3 | NP_031611.1 | 5 - 6 | 433 | 104 | |
GenBank mRNA | AK012563.1 | - | 6 - 7 | 435 | 104 | |
AK088645.1 | - | 5 - 6 | 422 | 104 | ||
AK146096.1 | - | 5 - 6 | 421 | 104 | ||
AK152561.1 | - | 5 - 6 | 421 | 104 | ||
AK160746.1 | - | 5 - 6 | 421 | 104 | ||
AK164507.1 | - | 5 - 6 | 426 | 104 | ||
AK165888.1 | - | 5 - 6 | 421 | 104 | ||
AK167894.1 | - | 5 - 6 | 420 | 104 | ||
AK168083.1 | - | 5 - 6 | 421 | 104 | ||
BC003327.1 | - | 5 - 6 | 433 | 3156634 | 104 | |
BC004659.1 | - | 1 - 2 | 1329 | 3497991 | 104 | |
BC005763.1 | - | 5 - 6 | 407 | 2655513 | 104 | |
D10024.1 | - | 4 - 5 | 366 | 104 | ||
M14044.1 | - | 4 - 5 | 366 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |