Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mre11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018736.2 | NP_061206.1 | 11 - 12 | 1355 | 91 | |
GenBank mRNA | AK041248.1 | - | 11 - 12 | 1354 | 91 | |
AK141214.1 | - | 11 - 12 | 1367 | 91 | ||
AK146234.1 | - | 10 - 11 | 1307 | 91 | ||
AK146370.1 | - | 10 - 11 | 1307 | 91 | ||
AK168180.1 | - | 10 - 11 | 1307 | 91 | ||
AK168208.1 | - | 11 - 12 | 1390 | 91 | ||
BC065144.1 | - | 11 - 12 | 1355 | 30627207 | 91 | |
U58987.1 | - | 11 - 12 | 1390 | 91 | ||
U60318.1 | - | 10 - 11 | 1309 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |