Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285853.1 | NP_001272782.1 | 2 - 3 | 337 | 97 | |
GenBank mRNA | AK018182.1 | - | 2 - 3 | 189 | 97 | |
AK079816.1 | - | 2 - 3 | 130 | 97 | ||
AK158883.1 | - | 2 - 3 | 195 | 97 | ||
S77473.1 | - | 2 - 3 | 149 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Mus musculus Probe spans exons |