Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161483.1 | NP_001154955.1 | 7 - 8 | 1082 | 73 | |
GenBank mRNA | AK086574.1 | - | 3 - 4 | 586 | 73 | |
BC056377.1 | - | 7 - 8 | 1075 | 6850243 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |