Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020626.2 | NP_065651.1 | 2 - 3 | 201 | 112 | |
GenBank mRNA | AF178085.1 | - | 2 - 3 | 201 | 112 | |
AK002337.1 | - | 2 - 3 | 147 | 112 | ||
AK148534.1 | - | 2 - 3 | 198 | 112 | ||
BC049912.1 | - | 2 - 3 | 123 | 4973401 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |