Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170745.1 | NP_001164216.1 | 6 - 7 | 1174 | 116 | |
NM_001170746.1 | NP_001164217.1 | 7 - 8 | 1339 | 116 | ||
NM_015823.3 | NP_056638.1 | 6 - 7 | 1174 | 116 | ||
GenBank mRNA | AK039336.1 | - | 3 - 4 | 159 | 116 | |
AK122349.1 | - | 5 - 6 | 634 | 116 | ||
AK147530.1 | - | 6 - 7 | 1175 | 116 | ||
BC059005.1 | - | 6 - 7 | 831 | 6847238 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |