Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177707.3 | NP_808375.1 | 2 - 3 | 431 | 72 | |
GenBank mRNA | AK036516.1 | - | 2 - 3 | 432 | 72 | |
BC067208.1 | - | 2 - 3 | 211 | 30286602 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |