Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009284.2 | NP_033310.2 | 3 - 4 | 540 | 68 | |
GenBank mRNA | AF481808.1 | - | 2 - 3 | 255 | 68 | |
AF481809.1 | - | 2 - 3 | 255 | 68 | ||
AK149563.1 | - | 3 - 4 | 500 | 68 | ||
AK163053.1 | - | 3 - 4 | 541 | 68 | ||
BC127054.1 | - | 3 - 4 | 330 | 40109712 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |