Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019467.2 | NP_062340.1 | 2 - 3 | 165 | 67 | |
GenBank mRNA | AK006184.1 | - | 2 - 3 | 165 | 67 | |
AK006562.1 | - | 2 - 3 | 166 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |