Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019697.3 | NP_062671.1 | 2 - 3 | 1469 | 66 | |
GenBank mRNA | AB045326.1 | - | 2075 | 66 | ||
AB093280.1 | - | 2 - 3 | 1950 | 66 | ||
AF107780.1 | - | 2 - 3 | 1279 | 66 | ||
AK032268.1 | - | 2260 | 66 | |||
AK032772.1 | - | 1 - 2 | 45 | 66 | ||
BC079667.1 | - | 2 - 3 | 1469 | 30356567 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |