Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011046 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
684489 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
624521 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891763 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
672137 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
722972 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
93108 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
859976 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
194214 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
508888 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
152724 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
587328 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906479 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900434 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
620547 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45401 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360737 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
315538 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
399040 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
441029 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
597942 |
AK042798.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
597942 |
NM_009399.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
75100 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
892618 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183780 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
193142 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
550505 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
737718 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
964928 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563797 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
477978 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426555 |
AK031970.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426555 |
AK173294.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426555 |
BC076582.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426555 |
BC079588.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426555 |
BC094219.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
426555 |
NM_183141.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
784956 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
140970 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
578979 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876951 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
313542 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
176952 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
710126 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
940600 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347495 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
361258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
773521 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
268656 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656992 |
AK148122.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656992 |
BC006796.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656992 |
BC085501.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656992 |
NM_008841.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656992 |
Y13569.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
952237 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
477612 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
291740 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
536637 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600051 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
410796 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306514 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
230653 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543409 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
645106 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
204021 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
93520 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
945850 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3513 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
224333 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
28596 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
848027 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
203678 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379311 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
201456 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
102600 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
109170 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65291 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
131483 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
116843 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52418 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111414 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
116728 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11867 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
162490 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
51437 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
1074 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
18942 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
172941 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
178071 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P535430 |
AY243472.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P535430 |
NR_024051.1 |
mouse_transcriptome |