Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080446.2 | NP_536694.2 | 1 - 2 | 276 | 122 | |
GenBank mRNA | AB048542.1 | - | 1 - 2 | 201 | 122 | |
AK043127.1 | - | 1 - 2 | 265 | 122 | ||
AK131122.1 | - | 1 - 2 | 298 | 122 | ||
BC068140.1 | - | 1 - 2 | 204 | 6843107 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |