Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167777.1 | NP_001161249.1 | 8 - 9 | 829 | 98 | |
GenBank mRNA | AK082659.1 | - | 8 - 9 | 829 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |