Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289546.1 | NP_001276475.1 | 2 - 3 | 231 | 90 | |
NM_175007.2 | NP_778172.1 | 2 - 3 | 231 | 90 | ||
GenBank mRNA | AK047144.1 | - | 2 - 3 | 185 | 90 | |
AK132168.1 | - | 2 - 3 | 232 | 90 | ||
BC054718.1 | - | 2 - 3 | 211 | 5686478 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |