Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700018081.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018081.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018081.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018079.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018079.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018079.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018086.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018086.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018086.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018085.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018085.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018085.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018077.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018077.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018077.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018082.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018082.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018082.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018072.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018072.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018072.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018090.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018090.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018090.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018091.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018091.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018091.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018073.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018073.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018073.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018084.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018084.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018084.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018074.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018074.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018074.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018083.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018083.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018083.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018087.mm.1 |
TC0700002044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018087.mm.1 |
NM_001081118.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018087.mm.1 |
NM_001081118.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018080.mm.1 |
TC0700002044.mm.1 |
MTA TC |